Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 NS1 Antibody (HL2130) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325526
Description
Influenza A H1N1 NS1 Monoclonal antibody specifically detects Influenza A H1N1 NS1 in Virus samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Influenza A H1N1 NS1 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, Immunocytochemistry | |
HL2130 | |
Western Blot 1:500-1:3000, Immunocytochemistry/ Immunofluorescence | |
Influenza NS1 | |
Recombinant fragment of Influenza A H1N1 NS1. | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction