Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody (2F205), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP224871AF594

Catalog No. NP224871A94


Only null left
Add to Cart

Description

Description

Influenza A nucleoprotein Monoclonal specifically detects Influenza A nucleoprotein in Virus samples. It is validated for ELISA.
Specifications

Specifications

Influenza A nucleoprotein
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
Mouse
Protein G purified
Immunology
Virus
IgG1 κ
ELISA
2F205
ELISA
Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Recombinant influenza A protein was used as the immunogen
0.1 mL
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.