Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody (2F205) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$181.72 - $420.02

Specifications

Antigen Influenza A nucleoprotein
Clone 2F205
Concentration 1.0 mg/mL
Dilution ELISA 0.1- 2.0 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224871SS
SDP
View Documents
Novus Biologicals
NBP224871SS
0.025 mg
Each for $181.72
Only null left
Add to Cart
 
NBP224871
SDP
View Documents
Novus Biologicals
NBP224871
0.1 mg
Each for $420.02
Only null left
Add to Cart
 
Description

Description

Influenza A nucleoprotein Monoclonal specifically detects Influenza A nucleoprotein in Virus samples. It is validated for ELISA.
Specifications

Specifications

Influenza A nucleoprotein
1.0 mg/mL
ELISA
Unconjugated
Mouse
Virus
Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Recombinant influenza A protein was used as the immunogen
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2F205
ELISA 0.1- 2.0 ug/ml
Monoclonal
Purified
RUO
O73619
394335
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.