Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 PA Antibody - (A/Puerto Rico/8/1934), Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP242876

Catalog No. NBP242876


Only null left
Add to Cart

Description

Description

Specifically detects Influenza A Virus PA in Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

Influenza A H1N1 PA Polyclonal antibody specifically detects Influenza A H1N1 PA in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Influenza A H1N1 PA
Polyclonal
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Influenza PA
Recombinant protein encompassing a sequence within the C-terminus region of Influenza A virus PA (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Affinity Purified
RUO
Virus
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Rabbit
83 kDa
0.1 mL
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only