Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A (H1N1) virus NA Mouse anti-Influenza virus, Clone: 3F11D7, Abnova™

Catalog No. 89035659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-659 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-659 Supplier Abnova Corporation Supplier No. MAB14223

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant influenza A (H1N1) virus NA.

  • High specificity
  • High reactivity

Specifications

Antigen Influenza A (H1N1) virus NA
Applications Western Blot
Classification Monoclonal
Clone 3F11D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant influenza A (H1N1) virus NA.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.5 mg/mL PBS, pH 7.4 (0.02% sodium azide).
Host Species Mouse
Immunogen Recombinant protein corresponding to full length influenza A (H1N1) 2009 virus NA.
Purification Method Protein A purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus
Content And Storage Store at -20°C or lower.
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL.
The reconstituted antibody can be stored for 2-3 weeks at 4°C. For long term storage, aliquot and st
Form Lyophilized
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.