Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H3N2 Hemagglutinin Antibody - (A/Texas/1/77) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017606 0.1 mL
NB10064556 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017606 Supplier Novus Biologicals Supplier No. NB100645560.1ML
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Influenza A H3N2/HA Polyclonal specifically detects Influenza A H3N2/HA in Influenza A Virus H3N2 samples. It is validated for Immunocytochemistry/Immunofluorescence, Haemagglutination.

Specifications

Antigen Influenza A H3N2
Applications Hemagglutination Assay, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000, Haemagglutination
Formulation PBS with 0.09% Sodium Azide
Host Species Goat
Immunogen Influenza A, Texas 1/77 strain (H3N2).
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for Influenza A H3N2 by Haemagglutination inhibition. Does not react with influenza B, RSV, Para 1-3 or Adenovirus, but may react with proteins present on uninfected chicken cells.
Target Species Influenza virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.