Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H3N2 M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA17223
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA17223 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA17223 Supplier Invitrogen™ Supplier No. PA17223
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PA1-7223 detects the Influenza A virus. PA1-7223 has been successfully used in immunofluorescence procedures. The PA1-7223 immunogen is influenza A, Phillipines (H3N2). Recommended short term storage (up to 6 months) is at 4ºC. For longer storage, aliquot and freeze.

Influenza A virus is an enveloped virus encoding 10 polypeptides. RNA segment 7 encodes for two proteins: M1 (matrix 1) and M2 (matrix 2). M1 protein is encoded by an mRNA that is colinear, while M2 protein is synthesized from spliced mRNA. M2 protein is a transmembrane protein composed of three Domains: 1) 24 residues representing the N-terminal region, 2) 19 hydro-phobic residues that serve as a membrane anchor, and 3) 54 residues near the C-terminal in the cytoplasmic domain. The M2 protein has been found to play a role in Influenza replication and assembly of virion particles. Further experimentation has demonstrated that this protein is an acid-activated ion channel for virus replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H3N2 M1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias H3N2; H3N2 M1; H3N2 Matrix protein; Influenza H3N2
Host Species Goat
Immunogen Influenza A, Phillipines (H3N2).
Purification Method IgG Fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.