Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H3N2 Virions Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PA173047
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173047 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173047 Supplier Invitrogen™ Supplier No. PA173047
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73047 antibody reacts with Influenza A virions H3N2. PA1-73047 has been successfully used in Immunofluorescence applications. The PA1-73047 antibody was raised against Influenza A, strain Texas 1/77 (H3N2).

Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H3N2 Virions
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias H3N2; H3N2 virions; Influenza H3N2
Host Species Goat
Immunogen Influenza A, strain Texas 1/77 (H3N2).
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.