Learn More
Invitrogen™ Influenza A H4N6 HA Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for H4N6 (A/mallard/Ohio/657/2002) HA/Hemagglutinin.
Specifications
Specifications
| Antigen | Influenza A H4N6 HA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | flu hemagglutinin; influenza hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Recombinant H4N6 (A/mallard/Ohio/657/2002) HA protein. |
| Purification Method | Antigen affinity chromatography, Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.