Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H4N6 NA (A/mallard/Ohio/657/2002) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA581751
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H4N6 (A/mallard/Ohio/657/2002) Neuraminidase/NA.

Specifications
Influenza A H4N6 NA (A/mallard/Ohio/657/2002) | |
Polyclonal | |
PBS with no preservative | |
Recombit Influenza A H4N6 (A/mallard/Ohio/657/2002) Neuraminidase/NA Protein | |
100 μL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
ELISA | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction