Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H5 Monoclonal Antibody (D86J)
GREENER_CHOICE

Catalog No. PIMA518212
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518212 Supplier Invitrogen™ Supplier No. MA518212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific for the H5 protein and does not cross-react with other Influenza H types. Purity is >90%.

Haemagglutinin is an antigenic glycoprotein which allows viral attachment and entry into the cell. Sixteen subtypes of haemagglutinin (H1-H16) have been described, of which H1, H2 and H3 infect humans. H5 and H7 normally result in avian disease, with some highly pathogenic H5N1 strains causing 100% mortality in poultry. H5N1 has in some cases mutated to infect humans, with 60% mortality. Haemagglutinin H5 is from the Influenza A virus H5N1. Influenza A belongs to the Orthomyxoviridae family, and is a negative sense single-stranded RNA virus which results in respiratory disease. Haemagglutinin is one of the most medically relevant antigens on influenza as it is a target for antiviral drugs and antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H5
Applications ELISA
Classification Monoclonal
Clone D86J
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Influenza H5; Influenza NP-A
Host Species Mouse
Immunogen Influenza A virus native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.