Learn More
Invitrogen™ Influenza A H5N1 HA Monoclonal Antibody

Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Influenza A H5N1 HA |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1D10 |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | flu hemagglutinin; influenza hemagglutinin |
| Host Species | Mouse |
| Immunogen | Influenza A virus H5N1 HA (Hemagglutinin) (A/Hatay/2004/(H5N1)). The exact sequence is proprietary. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.