Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (1E7D8), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP175494PCP
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza A H5N1 Hemagglutinin | |
Monoclonal | |
PerCP | |
PBS | |
A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Western Blot, ELISA | |
1E7D8 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction