Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (2B7), FITC - (A/Vietnam/HN31242/2007), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222605F
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza A Virus H5N1 samples. It is validated for ELISA.Specifications
Influenza A H5N1 Hemagglutinin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HA | |
Recombinant Influenza A H5N1 Hemagglutinin (17-338aa) purified from Baculovirus | |
0.1 mL | |
Primary | |
Anti-human H5N1/HA1 mAb, clone 2B7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human H5N1/HA1 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA | |
2B7 | |
ELISA | |
HA, Hemagglutinin | |
Mouse | |
Protein G purified | |
RUO | |
3654620 | |
Influenza virus, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction