Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (4E10C10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP175504
Description
Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.Specifications
Influenza A Hemagglutinin | |
Monoclonal | |
1 mg/mL | |
Western Blot 2.5 ug/ml, ELISA 2 ug/mL | |
HA epitope tag, HA1, HA2, Hemagglutinin | |
Mouse | |
Affinity Purified | |
RUO | |
Virus | |
IgG1, IgG2b |
Western Blot, ELISA | |
4E10C10 | |
Unconjugated | |
AAT76166 | |
HA | |
Recombinant protein corresponding to amino acids 17 - 338 of H5 HA1. | |
0.1 mg | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction