Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Rabbit anti-Influenza virus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$297.50 - $527.00
Specifications
Antigen | Influenza A H5N1 Hemagglutinin |
---|---|
Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza virus samples. It is validated for Western Blot, ELISASpecifications
Influenza A H5N1 Hemagglutinin | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Immunology, Virology Bacteria and Parasites | |
0.2μm filtered solution in PBS without Preservative | |
IgG | |
Protein A purified |
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Influenza virus | |
Purified, recombinant Influenza A H5N1 Hemagglutinin (ABD73284.1; Met1-Gln531) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title