Learn More
Invitrogen™ Influenza A H5N1 NA (A/Hubei/1/2010) Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H5N1 (A/Hubei/1/2010) Neuraminidase/NA.
Specifications
Specifications
| Antigen | Influenza A H5N1 NA (A/Hubei/1/2010) |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Avian H5N1; H5N1; Influenza A Neuraminidase |
| Host Species | Rabbit |
| Immunogen | Influenza A H5N1 (A/Hubei/1/2010) Neuraminidase/NA. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.