Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H5N8 NP (A/Astrakhan/3212/2020) Recombinant Rabbit Monoclonal Antibody (HL1323)
GREENER_CHOICE

Catalog No. PIMA547001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547001 Supplier Invitrogen™ Supplier No. MA547001
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

H5N8 is a highly pathogenic avian influenza (HPAI) originally identified in chinese domestic ducks The subtype H5N8 virus bears an H5N1 backbone and polymerase basic (PB) protein 1 (PB1), nucleoprotein (NP), and neuraminidase (NA) genes from non-H5N1 virus [PMID: 28117031].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H5N8 NP (A/Astrakhan/3212/2020)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1323
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias H5N8 NP; influenza H5N8
Host Species Rabbit
Immunogen Recombinant fragment corresponding to C-terminal region of Influenza A virus H5N8 Nucleoprotein (A/Astrakhan/3212/2020)). The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.