Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H6N1 Hemagglutinin Antibody (8A10D8), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306474AF700
Description
Influenza A H6N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H6N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza A H6N1 Hemagglutinin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H6N1 Hemagglutinin. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
8A10D8 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction