Learn More
Invitrogen™ Influenza A H6N1 MP1 Polyclonal Antibody

Description
This sequence is 100% conserved in chicken, duck, and porcine Influenza A.
Specifications
Specifications
| Antigen | Influenza A H6N1 MP1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene Alias | H6N1 MP1; Influenza H6N1 |
| Host Species | Rabbit |
| Immunogen | Amino acids 9-28 of avian flu matrix protein 1. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.