Learn More
Influenza A H7N1 Antibody (InA331), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP150815B
Catalog No. NBP150815B
Only null left
Add to Cart
Description
Description
Influenza A H7N1 Monoclonal specifically detects Influenza A H7N1 in Virus samples. It is validated for ELISA, Immunoassay.
Specifications
Specifications
Influenza A H7N1
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7).
0.25 mL
Primary
Store at 4C in the dark.
IgG1
ELISA, Immunoassay
InA331
ELISA, Immunoassay
Mouse
Protein G purified
RUO
Human, Virus
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.