Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H7N1 Antibody (InA334), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP150816AF532
Description
Influenza A H7N1 Monoclonal antibody specifically detects Influenza A H7N1 in Virus samples. It is validated for ELISA, ELISA, Direct ELISASpecifications
Influenza A H7N1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7). | |
0.25 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Sandwich ELISA, Direct ELISA | |
InA334 | |
ELISA, Sandwich ELISA, Direct ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction