Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza A H7N7 HA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534922
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifications
Influenza A H7N7 HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H7N7 HA; H7N7 hemagglutinin; Influenza H7N7; influenza hemagglutinin | |
Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus H7N7 HA (Hemagglutinin) (A/duck/Taiwan/33/1993(H7N7)). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.98 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction