Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H7N7 Hemagglutinin Antibody (19), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289790F
Description
Influenza A H7N7 Hemagglutinin Monoclonal antibody specifically detects Influenza A H7N7 Hemagglutinin in Virus samples. It is validated for ELISASpecifications
Influenza A H7N7 Hemagglutinin | |
Monoclonal | |
FITC | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
ELISA | |
19 | |
PBS | |
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A Hemagglutinin H7N7 protein. | |
0.2 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction