Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H7N7 Neuraminidase Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314650MFV450
Description
Influenza A H7N7 Neuraminidase Polyclonal antibody specifically detects Influenza A H7N7 Neuraminidase in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza A H7N7 Neuraminidase | |
| Polyclonal | |
| Western Blot, ELISA | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Influenza A H7N7 Neuraminidase (Accession#: AAR11367.1; His36-Ser471). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction