Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H7N9 HA (A/Anhui/1/2013) Monoclonal Antibody (06)
GREENER_CHOICE

Catalog No. PIMA529941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529941 Supplier Invitrogen™ Supplier No. MA529941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin/HA.

Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H7N9 HA (A/Anhui/1/2013)
Applications ELISA
Classification Monoclonal
Clone 06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Mouse
Immunogen Recombinant Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin/HA Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.