Learn More
Invitrogen™ Influenza A H9N2 NA (A/Hong Kong/1073/99) Monoclonal Antibody (02)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase/NA.
Specifications
Specifications
| Antigen | Influenza A H9N2 NA (A/Hong Kong/1073/99) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 02 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Host Species | Mouse |
| Immunogen | Recombinant Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase/NA Protein. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.