Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A HA Human anti-Virus, Clone: rFluA-20, MilliporeSigma™

Catalog No. MABF268525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF268525 25 μg
MABF2685100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF268525 Supplier MilliporeSigma Supplier No. MABF268525UG
Add to Cart
Add to Cart

Human Monoclonal Antibody

Influenza A HA Monoclonal antibody specifically detects Influenza A HA in Virus samples. It is validated for ELISA, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A HA
Applications ELISA, Neutralization
Classification Monoclonal
Clone rFluA-20
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified human recombinant monoclonal antibody IgG1 in PBS without azide.
Host Species Human
Immunogen PBMCs from human donor, who was subjected to Flu vaccine multiple times, were immortalized and used generate human hybridoma cell lines secreting human mAbs.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.