Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A HA Recombinant Rabbit Monoclonal Antibody (2)
GREENER_CHOICE

Catalog No. PIMA530006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530006 Supplier Invitrogen™ Supplier No. MA530006
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for H1N1 (A/California/04/2009) HA H1N1 (A/California/07/2009) HA.

Infectious diseases are the leading cause of death worldwide. AIDS, tuberculosis (TB), malaria, diarrhoeal and respiratory infections account for 78% of deaths caused by infectious disease. As many infectious diseases are controlled, new diseases emerge and old diseases become resistant to current drugs. Many infectious diseases have been associated with an increase risk of carcinoma. Influenza continues to attract researchers as new strains appear by the ability of the influenza gene to mix with different forms of the virus. Recently, research on SARS and West Nile virus has risen due to the increased number of infections. These antibodies assist in research by detecting the infectious disease agent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A HA
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Rabbit
Immunogen Recombinant H1N1 HA protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.