Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A Hemagglutinin H1 Antibody (InA97), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073158UV
Description
Influenza A Hemagglutinin H1 Monoclonal antibody specifically detects Influenza A Hemagglutinin H1 in Virus samples. It is validated for Western Blot, ELISA.Specifications
Influenza A Hemagglutinin H1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Detects H1 of Influenza A as well as recombinant H1 with high specificity and does not interact with haemagglutinin H3. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
InA97 | |
Western Blot, ELISA, Sandwich ELISA | |
HA, Hemagglutinin, Hemagglutinin HA | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/NewCaledonia/20/99 (strain H1N1). | |
0.2 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction