Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A Hemagglutinin H3 Antibody (InA246), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073162
Description
Influenza A Hemagglutinin H3 Monoclonal antibody specifically detects Influenza A Hemagglutinin H3 in Virus samples. It is validated for Western Blot, ELISA.Specifications
Influenza A Hemagglutinin H3 | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A purified | |
RUO | |
Influenza A haemagglutinin H3. | |
Store at 4C. Do not freeze. | |
IgG2a |
Western Blot, ELISA | |
InA246 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Shangdong/9/93 (strain H3N2). | |
0.2 mg | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction