Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A M1 Antibody (1G1A12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250480F
Description
Influenza A M1 Monoclonal antibody specifically detects Influenza A M1 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.Specifications
Influenza A M1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
GST-Influenza A M1 fusion protein | |
0.1 mL | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
1G1A12 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction