Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A M2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532233 Supplier Invitrogen™ Supplier No. PA532233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Influenza virus (A/WSN/1933(H1N1))-infected DF-1 cell lysates, H1N1-infected H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Influenza A virus is an enveloped virus encoding 10 polypeptides. RNA segment 7 encodes for two proteins: M1 (matrix 1) and M2 (matrix 2). M1 protein is encoded by an mRNA that is colinear, while M2 protein is synthesized from spliced mRNA. M2 protein is a transmembrane protein composed of three Domains: 1) 24 residues representing the N-terminal region, 2) 19 hydro-phobic residues that serve as a membrane anchor, and 3) 54 residues near the C-terminal in the cytoplasmic domain. The M2 protein has been found to play a role in Influenza replication and assembly of virion particles. Further experimentation has demonstrated that this protein is an acid-activated ion channel for virus replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A M2
Applications Immunomicroscopy, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias Influenza M2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Influenza A virus M2 (matrix protein) (A/Puerto Rico/8/1934(H1N1). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.