Learn More
Invitrogen™ Influenza A NP Monoclonal Antibody (9C11)

Description
MA5-17101 targets Influenza A virus Nucleoprotein in indirect ELISA and WB applications and shows reactivity with Virus samples. The MA5-17101 immunogen is purified recombinant fragment of Influenza A virus nucleoprotein expressed in E. Coli strain BL21(DE3).
Specifications
Specifications
| Antigen | Influenza A NP |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 9C11 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene Alias | Influenza nucleoprotein |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of Influenza A virus nucleoprotein expressed in E. Coli strain BL21(DE3). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.