Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza A NP Monoclonal Antibody (AA5H)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516291
Description
Influenza A NP Monoclonal Antibody for ICC/IF
The nucleoprotein (NP) of Influenza A virus encapsidates the negative strand of the viral RNA and is essential for replicative transcription. It may also be involved in other essential functions throughout the virus life cycle. As well as binding ssRNA, NP is able to self associate to form large oligomeric complexes. NP is able to interact with a variety of other macromolecules of both viral and cellular origins. It binds the PB1 and PB2 subunits of the polymerase and the matrix protein M1. NP has also been shown to interact with at least four cellular polypeptide families: nuclear import receptors of the importin class, filamentous (F) actin, the nuclear export receptor CRM1 and a DEAD box helicase BAT1/UAP56 (Portela et al 2002).
Specifications
Influenza A NP | |
Monoclonal | |
8.88 mg/mL | |
PBS with 15mM sodium azide; pH 7.5 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry | |
AA5H | |
Unconjugated | |
Influenza nucleoprotein | |
A/Puerto Rico/8/34 (H1N1) and A/Bangkok/1/79 (H3N2) virus. | |
2 mg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction