Learn More
Invitrogen™ Influenza A NP Polyclonal Antibody

Description
Suggested positive control: recombinant Influenza A.
Specifications
Specifications
| Antigen | Influenza A NP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene Alias | Influenza nucleoprotein |
| Host Species | Rabbit |
| Immunogen | Recombinant Influenza A H1N1 Nucleoprotein. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.