Learn More
Invitrogen™ Influenza A NS1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 88% homologous in Chicken, Bovine, Opossum, Monkey and 83% homologous in Canine.
Specifications
Specifications
| Antigen | Influenza A NS1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene Alias | Influenza NS1 |
| Host Species | Rabbit |
| Immunogen | Within the range of amino acids 24-84 of human Influenza A H1N1 nonstructural protein. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.