Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A NS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532235 Supplier Invitrogen™ Supplier No. PA532235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Influenza virus (A/WSN/1933(H1N1))-infected DF-1 cell lysates. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years. Two distinct non-structural proteins NS1 and NS2 are translated from one of the eight RNA segments by using different reading frames.The influenza A virus NS2 is located in the nucleus and the cytoplasm. NS2 is essential for nuclear export of influenza virus ribonucleoprotein (RNP) complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A NS2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias Influenza NS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus NS2 (nonstructural protein) (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.