Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A nucleoprotein Antibody (010), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312741JF549
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Influenza A virus H3N2 samples. It is validated for Western Blot, ELISASpecifications
Influenza A nucleoprotein | |
Monoclonal | |
Janelia Fluor 549 | |
Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (Uniprot#: P22435.2; Met1-Asn498). | |
100 μg | |
Immunology | |
Influenza A virus H3N2 | |
Purified |
Western Blot, ELISA | |
010 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction