Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A nucleoprotein Antibody (1331), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065046B
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Ferret, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
Influenza A nucleoprotein | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
NP | |
From Texas strain | |
0.1 mL | |
Primary | |
Ferret, Virus | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1331 | |
ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Flu | |
Mouse | |
Protein A purified | |
RUO | |
Specific for the nucleoprotein of numerous strains and hence gives a 'nuclear' staining in IF. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction