Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A nucleoprotein Antibody (1331) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017686 0.025 mg
NB10065046 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017686 Supplier Novus Biologicals Supplier No. NB100650460.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A Virus Monoclonal specifically detects Influenza A Virus in Ferret, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Influenza A Virus
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 1331
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:200-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:100-1:500
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P18072
Gene Alias Flu
Gene Symbols NP
Host Species Mouse
Immunogen From Texas strain
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the nucleoprotein of numerous strains and hence gives a 'nuclear' staining in IF.
Target Species Virus, Ferret
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.