Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A nucleoprotein Rabbit anti-Virus, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABF182025UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF1820-25UL 25 μL
ABF1820-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABF1820-25UL Supplier MilliporeSigma Supplier No. ABF182025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Influenza A nucleoprotein, Cat. No. ABF1820, is a rabbit polyclonal antibody that detects influenza virus nucleoprotein and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A nucleoprotein
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum in 0.05% sodium azide.
Gene Alias Nucleoprotein;Nucleocapsid protein;Protein N
Gene Symbols NP
Host Species Rabbit
Immunogen Hig-tagged full length recombinant nucleoprotein from Influenza A virus.
Molecular Weight of Antigen ∽56 kDa observed; 56.10 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) Translation: AAD12236
Test Specificity This rabbit polyclonal antibody detects influenza A virus nucleoprotein in virus infected cells.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.