Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza A PB1 Recombinant Rabbit Monoclonal Antibody (HL1715)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547153
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Influenza A virus contains eight single stranded, negative sense RNAs. Each RNA is associated with viral RNA-dependent RNA polymerase which consists of PA, PB1 and PB2 proteins. PB1 is the subunit involved in the catalytic activity of nucleotide polymerization and is involved in the initiation of transcription. Both PB1 and PB2 can be crosslinked to synthetic RNA with the 3' terminal sequence of viral RNA. PB1 and PB2 may also be involved in recognition of the promoter and/or replication origin on template viral RNA. PB2 is the cap 1-recognition protein, which binds to the cap structure of host mRNA. The cap structure then acts as a primer for transcription to produce viral mRNA.
Specifications
Influenza A PB1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot | |
HL1715 | |
Unconjugated | |
Influenza PB1 | |
Recombinant protein encompassing a sequence within the center region of Influenza A virus PB1 protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction