Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$188.70 - $474.50
Specifications
Antigen | Hemagglutinin/HA Peptide |
---|---|
Clone | 2908B |
Dilution | 1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry) |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11396100
![]() |
R&D Systems
MAB11396100 |
100 μg |
Each for $474.50
|
|
|||||
MAB11396SP
![]() |
R&D Systems
MAB11396SP |
25 μg |
Each for $188.70
|
|
|||||
Description
Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza virusN1 samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
Hemagglutinin/HA Peptide | |
1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry) | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2908B | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Influenza virus N1 | |
YP_009118626 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title