Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

influenza A virus Matrix 1 Mouse anti-Virus, Clone: 1G1A12, MilliporeSigma™

Catalog No. MABF2216100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2216100 100 μg
MABF221625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2216100 Supplier MilliporeSigma Supplier No. MABF2216100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

influenza A virus Matrix 1 Monoclonal antibody specifically detects influenza A virus Matrix 1 in Virus samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen influenza A virus Matrix 1
Applications Western Blot
Classification Monoclonal
Clone 1G1A12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen GST-tagged recombinant full-length Matrix 1 protein from Influenza A virus.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.