Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A virus nucleoprotein Mouse anti-Influenza virus, Clone: AA5H, Abnova™

Catalog No. 89035505 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-505 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-505 Supplier Abnova Corporation Supplier No. MAB14107

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Influenza A virus nucleoprotein.

  • High specificity
  • High reactivity

Specifications

Antigen Influenza A virus nucleoprotein
Applications Immunofluorescence
Classification Monoclonal
Clone AA5H
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Influenza A virus nucleoprotein.
Dilution Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (< 15 mM sodium azide).
Host Species Mouse
Immunogen A/Puerto Rico/8/34 (H1N1) and A/Bangkok/1/79 (H3N2) virus.
Purification Method Protein A affinity purification
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.