Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Antibody (1131 (2B2)) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100650340.025MG
Description
Influenza B Virus Monoclonal specifically detects Influenza B Virus in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Influenza B Virus | |
Monoclonal | |
1.0 mg/mL | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Mouse | |
Protein A purified | |
RUO | |
NB100-65034 exhibits nuclear staining in immunofluoresence and is therefore thought to recognize a nuclear protein. Influenza virus type B is a genus in the virus family Orthomyxoviridae responsible for acute respiratory illnesses in humans. It varies antigenically from influenza virus types A and C and is not divided into any subtypes. The gradual evolution of Influenza virus type B occurs via a series of mutations known as antigenic drift, in the surface glycoprotein haemagglutinin (HA), enabling the virus to evade the immune system and potentially re-infect the same host. HA glycoproteins from the Influenza virus type B are included in the annually updated trivalent flu vaccine, along with the Influenza virus type A subtypes H3N2 and H1N1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence | |
1131 (2B2) | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Influenza B virus preparation | |
0.025 mg | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction