Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Antibody (1131 (2B2)), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065034C
Description
Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
Influenza B Hemagglutinin 2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Influenza B virus preparation | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
1131 (2B2) | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Mouse | |
Protein A purified | |
RUO | |
NB100-65034 exhibits nuclear staining in immunofluoresence and is therefore thought to recognize a nuclear protein. Influenza virus type B is a genus in the virus family Orthomyxoviridae responsible for acute respiratory illnesses in humans. It varies antigenically from influenza virus types A and C and is not divided into any subtypes. The gradual evolution of Influenza virus type B occurs via a series of mutations known as antigenic drift, in the surface glycoprotein haemagglutinin (HA), enabling the virus to evade the immune system and potentially re-infect the same host. HA glycoproteins from the Influenza virus type B are included in the annually updated trivalent flu vaccine, along with the Influenza virus type A subtypes H3N2 and H1N1. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction