Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza B HA Monoclonal Antibody (B35G)
GREENER_CHOICE

Catalog No. PIMA516289
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516289 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516289 Supplier Invitrogen™ Supplier No. MA516289
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza B HA Monoclonal Antibody for ICC/IF, LF

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B HA
Applications Lateral Flow, Immunocytochemistry
Classification Monoclonal
Clone B35G
Concentration 6.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.5
Gene Alias flu hemagglutinin; Influenza B; influenza b hemagglutinin; Influenza HA; influenza hemagglutinin
Host Species Mouse
Immunogen B/Lee/40 and B/Singapore/-222/79 virus.
Purification Method Protein A
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.