Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Hemagglutinin Antibody (10H6C8G4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306569B
Description
Influenza B Hemagglutinin Monoclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza B Hemagglutinin | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
10H6C8G4 | |
Western Blot, ELISA | |
B1AG46 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinantInfluenza B Hemagglutinin (ACA33493.1; Met 1-Ala 555). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction